Tetrahydrofuran-d{8}, 99.5% (Isotopic), Thermo Scientific Chemicals
Tetrahydrofuran-d{8}, 99.5% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofuran-d{8}, 99.5% (Isotopic), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036551.03
also known as 036551-03
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.00 
Save 10.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.00 
Save 10.35 (15%)
Add to cart
Tetrahydrofuran-d{8}, 99.5% (Isotopic), Thermo Scientific Chemicals
Catalog number036551.03
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.00 
Save 10.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1693-74-9
IUPAC Name(²H₈)oxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-SVYQBANQSA-N
SMILES[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay from Supplier's CofA≥99.5% Isotopic enrichment by ¹H nmr
Assay from Supplier's CofA≥99.5% ¹H nmr
Water Content (Karl Fischer Titration)≤ 300ppm
Deuterated THF is used as a solvent in many organic reactions and in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only