Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036554.22
100 g, Each
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036554.22
100 g, Each
Quantity
Catalog number: 036554.22
also known as 036554-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-13-1
IUPAC Name
λ²-cobalt(2+) hexahydrate dichloride
Molecular Formula
Cl2CoH12O6
InChI Key
GFHNAMRJFCEERV-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Insoluble material
0.01%
Assay (unspecified)
98.0-102.0% Cl₂Co.6H₂O
Impurity content
Ca 0.005%
Impurity content
Cu 0.002%
Impurity content
Fe 0.005%

Description

Cobalt chloride is used as a humidity indicator in weather instruments. It finds use in electroplating of cobalt, in invisible ink, in organic chemistry and is a precursor to cobaltocene(bis (cyclopentadienyl) cobalt(II)).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text