Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 35% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 036560.36
500 g, Each
Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 35% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 036560.36
500 g, Each
Quantity
Catalog number: 036560.36
also known as 036560-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-98-5
IUPAC Name
tetraethylazanium hydroxide
Molecular Formula
C8H21NO
InChI Key
LRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES
[OH-].CC[N+](CC)(CC)CC
Identification (FTIR)
Conforms (non-U.S. specification)
Color (APHA)
≤100
Assay (Aqueous acid-base Titration)
≥32.5 to ≤37.5%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.4040-1.4070 @ 20°C (non-U.S. specification)

Description

Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text