Thermo Scientific Chemicals

Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene, Thermo Scientific Chemicals

Catalog number: 036563.AC
25 mL, Each
Thermo Scientific Chemicals

Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene, Thermo Scientific Chemicals

Catalog number: 036563.AC
25 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 036563.AC
also known as 036563-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1184-61-8
IUPAC Name
tetrakis(propan-2-ol) tin
Molecular Formula
C12H32O4Sn
InChI Key
VUDAJANAMZGVOP-UHFFFAOYSA-N
SMILES
[Sn].CC(C)O.CC(C)O.CC(C)O.CC(C)O
Appearance (Color)
Pale yellow
Assay from Supplier's CofA
≥97.5% (metals basis, neat)
Concentration (w/v)
9.5-10.5%
Form
Liquid

Description

Tin(IV) isopropoxide isopropanol is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, thin films doped tin oxide nanoparticles, thin films and are widely studied as gas sensors. Tin(IV) precursor solution was prepared from tin(IV) isopropoxide dissolved in isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text