Niobium isopropoxide, 99% (metals basis), 10% w/v in isopropanol/hexane (50:50), Thermo Scientific Chemicals
Niobium isopropoxide, 99% (metals basis), 10% w/v in isopropanol/hexane (50:50), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium isopropoxide, 99% (metals basis), 10% w/v in isopropanol/hexane (50:50), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number 036572.AE
also known as 036572-AE
Price (USD)/ Each
426.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
426.00
Add to cart
Niobium isopropoxide, 99% (metals basis), 10% w/v in isopropanol/hexane (50:50), Thermo Scientific Chemicals
Catalog number036572.AE
Price (USD)/ Each
426.00
-
Add to cart
Chemical Identifiers
CAS18368-80-4
IUPAC Namepentakis(propan-2-ol) niobium
Molecular FormulaC15H40NbO5
InChI KeyFTYCGXPVPAOYKN-UHFFFAOYSA-N
SMILES[Nb].CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤1.0%
FormLiquid
Niobium isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only