Thermo Scientific Chemicals

Gallium(III) isopropoxide, mixture of oligomers, 99%, Thermo Scientific Chemicals

Catalog number: 036583.06
5 g, Each
Thermo Scientific Chemicals

Gallium(III) isopropoxide, mixture of oligomers, 99%, Thermo Scientific Chemicals

Catalog number: 036583.06
5 g, Each
Quantity
Catalog number: 036583.06
also known as 036583-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4452-61-3
IUPAC Name
tris(propan-2-ol) gallium
Molecular Formula
C9H24GaO3
InChI Key
FUJNSPPLSODXIC-UHFFFAOYSA-N
SMILES
[Ga].CC(C)O.CC(C)O.CC(C)O
Form
Viscous liquid/solid
Appearance (Color)
Yellow
Assay from Supplier's CofA
≥98.5%
Chloride content
≤1.0%

Description

Gallium(III) isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text