Thermo Scientific Chemicals
Aluminum magnesium isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Catalog number: 036591.AC
25 mL, Each
Thermo Scientific Chemicals
Aluminum magnesium isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Catalog number: 036591.AC
25 mL, Each
Quantity
Catalog number: 036591.AC
also known as 036591-AC
Price (USD)
Quantity
-
Have Questions?
Description
Aluminum magnesium isopropoxide has been used to study the hydrolysis conditions on the properties of magnesium aluminate spinel powders.