Aluminum calcium isopropoxide, 99.8% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Aluminum calcium isopropoxide, 99.8% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum calcium isopropoxide, 99.8% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number 036594.AE
also known as 036594-AE
Price (USD)/ Each
278.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
278.00
Add to cart
Aluminum calcium isopropoxide, 99.8% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Catalog number036594.AE
Price (USD)/ Each
278.00
-
Add to cart
Specifications
Chemical Name or MaterialAluminum calcium isopropoxide
CAS23275-27-6
Health Hazard 1H225-H302-H314-H335-H336
Health Hazard 2GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only