Thermo Scientific Chemicals

Zinc telluride, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 036595.09
10 g, Each
Thermo Scientific Chemicals

Zinc telluride, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 036595.09
10 g, Each
Quantity
Catalog number: 036595.09
also known as 036595-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1315-11-3
Total Metal Impurities
0.002% max.

Description

Zinc telluride is a semiconductor material used in optoelectronics. It is used in blue LEDs, laser diodes, solar cells, components of microwave generators, electro-optic detector, holographic interferometry and laser optical phase conjugation devices. It is associated with lithium niobate and used for the generation of pulsed terahertz radiation in time-domain terahertz spectroscopy and terahertz imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text