Soda lime, ACS, Thermo Scientific Chemicals
Soda lime, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Soda lime, ACS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 036596.36
also known as 036596-36
Price (USD)/ Each
58.65
Online exclusive
65.30 
Save 6.65 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
65.30 
Save 6.65 (10%)
Add to cart
Soda lime, ACS, Thermo Scientific Chemicals
Catalog number036596.36
Price (USD)/ Each
58.65
Online exclusive
65.30 
Save 6.65 (10%)
-
Add to cart
Chemical Identifiers
CAS8006-28-8
IUPAC Namesodium oxocalcium hydroxide
Molecular FormulaCaHNaO2
InChI KeyHUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES[OH-].[Na+].O=[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤7.0% at 200°C
CommentCO₂ absorption capacity ≥19.0%
Particle SizeFines ≤1%
Soda lime's ability to absorb carbon dioxide makes it valuable in medical and surgical professions. It is also used as powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil CO2 efflux upon absorption. It is also useful for direct measurement of dissolved CO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only