Thermo Scientific Chemicals

Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals

Catalog number: 036602.36
500 g, Each
Thermo Scientific Chemicals

Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals

Catalog number: 036602.36
500 g, Each
Quantity
Catalog number: 036602.36
also known as 036602-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-66-6
IUPAC Name
zinc
Molecular Formula
Zn
InChI Key
HCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES
[Zn]
Impurity content
Pb: ≤ 0.01%
Particle Size
20 mesh
Appearance (Color)
Shiny grey
Form
Granules
Comment
Suitability for determination of As P.T.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text