Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiourea, ACS, 99% min., Thermo Scientific Chemicals

Reagent for organic synthesis
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 036609.18
also known as 036609-18
Price (USD)/ Each
36.65
Online exclusive
40.10 
Save 3.45 (9%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
36.65
Online exclusive
40.10 
Save 3.45 (9%)
Add to cart
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Catalog number036609.18
Price (USD)/ Each
36.65
Online exclusive
40.10 
Save 3.45 (9%)
-
Add to cart
Chemical Identifiers
CAS62-56-6
IUPAC Namethiourea
Molecular FormulaCH4N2S
InChI KeyUMGDCJDMYOKAJW-UHFFFAOYSA-N
SMILESNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Argentometric Titration)>/= 99.0% ( Dried basis)
Loss on Drying0.5% max
FormCrystalline powder
Melting Point174 - 177 deg C
Appearance (Color)White
View more
Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only