Diphenylamine, ACS, Thermo Scientific Chemicals
Diphenylamine, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylamine, ACS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036610.14
also known as 036610-14
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.50 
Save 3.85 (13%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.50 
Save 3.85 (13%)
Add to cart
Diphenylamine, ACS, Thermo Scientific Chemicals
Catalog number036610.14
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.50 
Save 3.85 (13%)
-
Add to cart
Chemical Identifiers
CAS122-39-4
IUPAC NameN-phenylaniline
Molecular FormulaC12H11N
InChI KeyDMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point52.5-54.0?
Impurity contentNO₃ P.T.
Residue on ignition0.03%
CommentSensitivity to nitrate P.T.
Solution Testin alcohol P.T.
Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only