Thermo Scientific Chemicals

Sodium sulfide nonahydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 036622.A1
1 kg, Each
Thermo Scientific Chemicals

Sodium sulfide nonahydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 036622.A1
1 kg, Each
Quantity
Catalog number: 036622.A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1313-84-4
IUPAC Name
disodium nonahydrate sulfanediide
Molecular Formula
H18Na2O9S
InChI Key
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Form
Crystals or powder or crystalline powder or lumps
Appearance (Color)
White or colorless to pale yellow
Assay (unspecified)
98.0% Na₂S.9H₂O min
Impurity content
Fe P.T.
Comment
May contain a small amount of water

Description

It is the precursor material for preparation of cadmium sulfide quantum dots. Also employed in the pulp and paper industry. It is likewise employed in the making of colors and dyestuffs. It is too useful in the trace quantities for the rapid synthesis of small silver nanocubes through mediating polyol reduction. It has as a significant role in breaking up the feather keratin useful for the preparation of bio-polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text