Thermo Scientific Chemicals

o-Tolidine dihydrochloride, ACS, Thermo Scientific Chemicals

Catalog number: 036626.22
100 g, Each
Thermo Scientific Chemicals

o-Tolidine dihydrochloride, ACS, Thermo Scientific Chemicals

Catalog number: 036626.22
100 g, Each
Quantity
Catalog number: 036626.22
also known as 036626-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
612-82-8
IUPAC Name
dihydrogen 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine dichloride
Molecular Formula
C14H18Cl2N2
InChI Key
LUKPNZHXJRJBAN-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1
Residue on ignition
0.1%
Comment
Sensitivity to chlorine P.T. (0.1ppm free chlorine)
Solution Test
P.T.

Description

o-Tolidine is employed as a colorimetric reagent for gold and free chlorine in water. Used in the synthesis of 9,9-bis(3-methyl-4-aminophenyl)fluorine. It is utilized for the detection of reducing sugars by thin layer chromatography. It is used in hair dyes, as a pharmaceutical intermediate and laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text