Cadmium chloride, anhydrous, ACS, 99.0% min, Thermo Scientific Chemicals
Cadmium chloride, anhydrous, ACS, 99.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium chloride, anhydrous, ACS, 99.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036629.36
also known as 036629-36
Price (USD)/ Each
326.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
384.00 
Save 57.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
326.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
384.00 
Save 57.35 (15%)
Add to cart
Cadmium chloride, anhydrous, ACS, 99.0% min, Thermo Scientific Chemicals
Catalog number036629.36
Price (USD)/ Each
326.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
384.00 
Save 57.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10108-64-2
IUPAC Namecadmium(2+) dichloride
Molecular FormulaCdCl2
InChI KeyYKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% CdCl₂ min
Impurity contentFe 0.001%
Impurity contentNH₄ 0.01%
Impurity contentPb 0.005%
Impurity contentZn 0.05%
View more
Cadmium chloride is a precursor material for the preparation of cadmium sulfide. It is mainly utilized in dyeing, pigments, photocopying and electroplating of cadmium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Preparation of cadmium sulfide, as analytical lab reagent, photography, manufacturing of special mirrors, ice-nucleating agent, lubricant, to absorb H2S in sulfide analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only