Thermo Scientific Chemicals

1-Propylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 036635.AK
250 mL, Each
Thermo Scientific Chemicals

1-Propylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 036635.AK
250 mL, Each
Quantity
Catalog number: 036635.AK
also known as 036635-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-10-8
IUPAC Name
propan-1-amine
Molecular Formula
C3H9N
InChI Key
WGYKZJWCGVVSQN-UHFFFAOYSA-N
SMILES
CCCN
Assay (unspecified)
≥99.0%
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

1-Propylamine is used in chemical analysis and to make other chemicals. It is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Also it can be used as petroleum additive and preservative. It is used in the production of fiber and engineering plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text