2-Chloroethanol, 99%, Thermo Scientific Chemicals
2-Chloroethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloroethanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
250 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036636.K2
Price (USD)/ Each
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS107-07-3
IUPAC Name2-chloroethan-1-ol
Molecular FormulaC2H5ClO
InChI KeySZIFAVKTNFCBPC-UHFFFAOYSA-N
SMILESOCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
2-Chloroethanol is used in the preparation of several dyes by the alkylation of aniline derivatives with chloroethanol. It is a building block in the production of pharmaceuticals and plasticizers. Also used for manufacture of thiodiglycol. It is a solvent for cellulose acetate and ethyl cellulose, textile printing dyes, in dewaxing, refining of rosin, extraction of pine lignin and the cleaning of machines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only