Thermo Scientific Chemicals

Isobutanol, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 036643.K7
4 L, Each
Thermo Scientific Chemicals

Isobutanol, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 036643.K7
4 L, Each
Quantity
Catalog number: 036643.K7
also known as 036643-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-83-1
IUPAC Name
2-methylpropan-1-ol
Molecular Formula
C4H10O
InChI Key
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILES
CC(C)CO
Free acid (titration)
0.0005meq/g
Solution Test
in water P.T.
Color (APHA)
10
Impurity content
Carbonyl compounds 0.01% (butyraldehyde) and 0.02% (2-butanone)
Residual water
0.1%

Description

Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text