Isobutanol, ACS, 99+%, Thermo Scientific Chemicals
Isobutanol, ACS, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isobutanol, ACS, 99+%, Thermo Scientific Chemicals

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.001%, Solubility in water P.T., Titratable acid 0.0005meq/g, H2O 0.1%, Carbonyl compounds 0.01% (butyraldehyde) and 0.02% (2-butanone)
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
500 mL
1 L
4 L
Catalog number 036643.M1
also known as 036643-M1
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
Add to cart
Isobutanol, ACS, 99+%, Thermo Scientific Chemicals
Catalog number036643.M1
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS78-83-1
IUPAC Name2-methylpropan-1-ol
Molecular FormulaC4H10O
InChI KeyZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILESCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)0.0005meq/g
Solution Testin water P.T.
Color (APHA)10
Impurity contentCarbonyl compounds 0.01% (butyraldehyde) and 0.02% (2-butanone)
Residual water0.1%
View more
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only