Thermo Scientific Chemicals

Glycerol, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 036646.AP
500 mL, Each
Thermo Scientific Chemicals

Glycerol, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 036646.AP
500 mL, Each
Quantity
Catalog number: 036646.AP
also known as 036646-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-81-5
IUPAC Name
propane-1,2,3-triol
Molecular Formula
C3H8O3
InChI Key
PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES
OCC(O)CO
Heavy Metals
(as Pb): ≤2ppm
Residue on ignition
≤0.005%
Impurity content
Acrolein and glucose: Passes Test
Impurity content
Chlorinated compounds (as Cl): ≤0.003%
Impurity content
Fatty acid esters (as butyric acid): ≤0.05%

Description

Glycerol is widely used in cosmetics, especially for production of soaps and lotions, solvents and emulsifiers. It may also be found in inks, adhesives and pharmaceuticals. It plays an important role in the manufacture of plastics and resins. Glycerol bonded with silica gel plays an important role for effective removal of traces of Co(II) from aqueous solutions. Also useful for optical clearing of gastric tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text