Thermo Scientific Chemicals

Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 036664.A3
2 kg, Each
Thermo Scientific Chemicals

Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 036664.A3
2 kg, Each
Quantity
Catalog number: 036664.A3
also known as 036664-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-92-9
IUPAC Name
2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Formula
C6H8O7
InChI Key
KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES
OC(=O)CC(O)(CC(O)=O)C(O)=O
Assay (Titration)
≥99.5%
Impurity content
Fe: ≤3ppm
Impurity content
Oxalate (C₂O₄): P.T. (limit about 0.05%)
Impurity content
Sulfur compounds (as SO₄): ≤0.002%
Chloride content
≤0.001%

Description

Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text