Thermo Scientific Chemicals

Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036665.22
100 g, Each
Thermo Scientific Chemicals

Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036665.22
100 g, Each
Quantity
Catalog number: 036665.22
also known as 036665-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5949-29-1
IUPAC Name
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular Formula
C6H10O8
InChI Key
YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Appearance (Color)
Colorless or white
Form
Solid
Assay (Aqueous acid-base Titration)
99.0-102.0 %
Residue on ignition
≤0.02%
Impurity content
Chloride ( Cl): ≤0.001%

Description

Citric acid monohydrate is used in the preparation of citrate buffer in platelets for intravital microscopy. It acts as a pH-control agent in foods, beverages and pharmaceuticals applications. It acts as an iron chelator. In animals, it improves the utilization of nutritional calcium. It is a useful buffer component for antigen and epitope unmasking. It is also used as an acidifier, a flavoring agent and a chelating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text