Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number 036665.A3
also known as 036665-A3
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
Add to cart
Citric acid monohydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
Catalog number036665.A3
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5949-29-1
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular FormulaC6H10O8
InChI KeyYASYEJJMZJALEJ-UHFFFAOYSA-N
SMILESO.OC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless or white
FormSolid
Assay (Aqueous acid-base Titration)99.0-102.0 %
Residue on ignition≤0.02%
Impurity contentChloride ( Cl): ≤0.001%
View more
Citric acid monohydrate is used in the preparation of citrate buffer in platelets for intravital microscopy. It acts as a pH-control agent in foods, beverages and pharmaceuticals applications. It acts as an iron chelator. In animals, it improves the utilization of nutritional calcium. It is a useful buffer component for antigen and epitope unmasking. It is also used as an acidifier, a flavoring agent and a chelating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only