Thermo Scientific Chemicals

Methyl Red sodium salt, ACS

Catalog number: 036667.22
100 g, Each
Thermo Scientific Chemicals

Methyl Red sodium salt, ACS

Catalog number: 036667.22
100 g, Each
Quantity
Catalog number: 036667.22
also known as 036667-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
845-10-3
IUPAC Name
sodium 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular Formula
C15H14N3NaO2
InChI Key
GNTPCYMJCJNRQB-UHFFFAOYSA-M
SMILES
[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
Comment
Clarity of alcohol solution P.T.
Comment
Clarity of aqueous solution P.T.
Comment
Visual transition interval from pH 4.2 (pink) to 6.2 (yellow)

Description

Methyl Red sodium salt is used as a pH indicator. In microbiology, it is used to make agar plates in bacteriology. Further, it is used as a coloring agent and also finds use in mouthwash, toothpaste and in cosmetics. It is also employed as a rocket propellant. In addition to this, it is used in the preparation of reversible stain for rapid detection of stained cells in each specimen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text