Thermo Scientific Chemicals

4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%, Thermo Scientific Chemicals

Catalog number: 036674.18
50 g, Each
Thermo Scientific Chemicals

4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%, Thermo Scientific Chemicals

Catalog number: 036674.18
50 g, Each
Quantity
Catalog number: 036674.18
also known as 036674-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55-55-0
IUPAC Name
bis(4-(methylamino)phenol); sulfuric acid
Molecular Formula
C14H20N2O6S
InChI Key
ZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
Form
Solid (discolors in air)
Comment
Suitability for determination of phosphate P.T.
Assay (Titration)
99.0-101.5%
Residue on ignition
<0.1%

Description

4-(Methylamino)phenol sulfate is a photographic developer used in black and white photography. It is also used as tone developers in developing agents. Further, it is used in oxidative hair dye formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text