Thermo Scientific Chemicals

Potassium permanganate, low in mercury, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036675.36
500 g, Each
Thermo Scientific Chemicals

Potassium permanganate, low in mercury, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036675.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 036675.36
also known as 036675-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7722-64-7
IUPAC Name
potassium manganesoylolate
Molecular Formula
KMnO4
InChI Key
VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES
[K+].[O-][Mn](=O)(=O)=O
Assay (unspecified)
99.0% KMnO₄ min
Impurity content
Hg 0.05ppm
Insoluble material
0.2%
Impurity content
Chloride and chlorate (as Cl) 0.005%
Impurity content
SO₄ 0.02%

Description

Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text