Thermo Scientific Chemicals

Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals

Catalog number: 036683.14
25 g, Each
Thermo Scientific Chemicals

Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals

Catalog number: 036683.14
25 g, Each
Quantity
Catalog number: 036683.14
also known as 036683-14
Price (USD)
Quantity
-
Have Questions?
Form
Powder
Assay (special)
Ni 50%
Appearance (Color)
Gray
Assay (special)
Fe 50%
Total Metal Impurities
≤ 0.5%

Description

Iron Nickel powder is used as an low expansion alloy for precision instruments, measuring tapes and weights

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text