Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 036683.36
also known as 036683-36
Price (USD)/ Each
229.65
Online exclusive
255.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
229.65
Online exclusive
255.00 
Save 25.35 (10%)
Add to cart
Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
Catalog number036683.36
Price (USD)/ Each
229.65
Online exclusive
255.00 
Save 25.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialIron Nickel powder
ColorSilver to Gray
Health Hazard 1H228-H317-H350-H372
Health Hazard 2Keep container tightly closed. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Take precautionary measures against static discharges. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
Health Hazard 3P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
View more
Iron Nickel powder is used as an low expansion alloy for precision instruments, measuring tapes and weights

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only