Thermo Scientific Chemicals

4-Dimethylaminobenzaldehyde, ACS, Thermo Scientific Chemicals

Catalog number: 036684.36
500 g, Each
Thermo Scientific Chemicals

4-Dimethylaminobenzaldehyde, ACS, Thermo Scientific Chemicals

Catalog number: 036684.36
500 g, Each
Quantity
Catalog number: 036684.36
also known as 036684-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-10-7
IUPAC Name
4-(dimethylamino)benzaldehyde
Molecular Formula
C9H11NO
InChI Key
BGNGWHSBYQYVRX-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=O)C=C1
Color (APHA)
Of alcohol solution: 60, max
Residue on ignition
0.1%
Solution Test
in alcohol P.T.
Appearance (Color)
Color of Hydrochloric acid solution: P.T.
Melting Point
73 TO 75 Deg C

Description

4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text