Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals
Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals

Form: approx. 9.1g/50x50mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 036691.22
also known as 036691-22
Price (USD)/ Each
79.65
Online exclusive
88.40 
Save 8.75 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
88.40 
Save 8.75 (10%)
Add to cart
Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals
Catalog number036691.22
Price (USD)/ Each
79.65
Online exclusive
88.40 
Save 8.75 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentAs 1ppm
Impurity contentPb 0.005%
Impurity contentCu 0.005%
Impurity contentFe 0.01%
Impurity contentSb 0.02%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only