Thermo Scientific Chemicals

Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals

Catalog number: 036694.K2
1 L, Each
Thermo Scientific Chemicals

Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals

Catalog number: 036694.K2
1 L, Each
Quantity
Catalog number: 036694.K2
also known as 036694-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10035-10-6
IUPAC Name
hydrogen bromide
Molecular Formula
BrH
InChI Key
CPELXLSAUQHCOX-UHFFFAOYSA-N
SMILES
Br
Assay (unspecified)
47.0-49.0% HBr
Heavy Metals
(ICP-OES) 5ppm
Impurity content
Fe 1ppm
Impurity content
Se 0.01ppm
Residue on ignition
0.002%

Description

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes, and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration, and dehydration, and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text