Thermo Scientific Chemicals

Calcium sulfate dihydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036700.30
250 g, Each
Thermo Scientific Chemicals

Calcium sulfate dihydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036700.30
250 g, Each
Quantity
Catalog number: 036700.30
also known as 036700-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10101-41-4
IUPAC Name
calcium dihydrate sulfate
Molecular Formula
CaH4O6S
InChI Key
PASHVRUKOFIRIK-UHFFFAOYSA-L
SMILES
O.O.[Ca++].[O-]S([O-])(=O)=O
Assay (unspecified)
98.0-102.0% CaSO₄.2H₂O
Impurity content
Cl 0.005% (≤50 ppm)
Impurity content
K 0.005% (≤50 ppm)
Impurity content
Mg 0.02% (≤200 ppm)
Impurity content
NO₃ Passes test

Description

Used as portland-cement retarder, tile and plaster. It is a source of sulfur and sulfuric acid. Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text