Thermo Scientific Chemicals

Lead(II) 2-ethylhexanoate, Thermo Scientific Chemicals

Catalog number: 036711.18
50 g, Each
Thermo Scientific Chemicals

Lead(II) 2-ethylhexanoate, Thermo Scientific Chemicals

Catalog number: 036711.18
50 g, Each
Quantity
Catalog number: 036711.18
also known as 036711-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
301-08-6
IUPAC Name
λ²-lead(2+) bis(2-ethylhexanoate)
Molecular Formula
C16H30O4Pb
InChI Key
RUCPTXWJYHGABR-UHFFFAOYNA-L
SMILES
[Pb++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Form
Viscous liquid
Assay (unspecified)
40.5 to 42.5% Pb

Description

Films of lead (II) 2-ethylhexanote can be cast on silicon solutions by spin coating. Lead (II) 2-ethylhexanote is a common solvent in metal-organic deposition (MOD) process. Intermetallic PtPb nanoparticles have been synthesized by the chemical reduction of dimethyl(1,5-cyclooctadiene)platinum and lead(II)2-ethylhexanoate by sodium naphthalide in THF or diglyme. The precursors used for the photochemical production of PZT or PLZT films were lead(II) 2-ethylhexanoate, lanthanum(III) 2-ethylhexanoate, zirconyl(IV) 2-ethylhexa- noate, and titanium(IV) isopropoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text