Thermo Scientific Chemicals

Sodium oxide, Thermo Scientific Chemicals

Catalog number: 036712.22
100 g, Each
Thermo Scientific Chemicals

Sodium oxide, Thermo Scientific Chemicals

Catalog number: 036712.22
100 g, Each
Quantity
Catalog number: 036712.22
also known as 036712-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-59-3
IUPAC Name
disodium oxidandiide
Molecular Formula
Na2O
InChI Key
KKCBUQHMOMHUOY-UHFFFAOYSA-N
SMILES
[O--].[Na+].[Na+]
Assay (Titration)
78% min

Description

Sodium oxide is used in chemical manufacturing, ceramics and glasses. Ethylbenzene dehydrogenation and carbon dioxide shift-reaction occurs using a sodium oxide/alumina catalyst. It is employed as catalyzing gasification of CO2 on carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text