Thermo Scientific Chemicals

3-Methyl-1-butanol, ACS, 98.5+%, Thermo Scientific Chemicals

Catalog number: 036716.K7
4 L, Each
Thermo Scientific Chemicals

3-Methyl-1-butanol, ACS, 98.5+%, Thermo Scientific Chemicals

Catalog number: 036716.K7
4 L, Each
Quantity
Catalog number: 036716.K7
also known as 036716-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-51-3
IUPAC Name
3-methylbutan-1-ol
Molecular Formula
C5H12O
InChI Key
PHTQWCKDNZKARW-UHFFFAOYSA-N
SMILES
CC(C)CCO
Form
Liquid
Residual water
≤0.5%
Impurity content
Acid and esters (as amyl acetate): ≤0.2%
Residue on Evaporation
≤0.003%
Free acid (titration)
≤0.002meq/g

Description

3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text