Thermo Scientific Chemicals

Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals

Catalog number: 036733.30
250 g, Each
Thermo Scientific Chemicals

Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals

Catalog number: 036733.30
250 g, Each
Quantity
Catalog number: 036733.30
also known as 036733-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
584-84-9
IUPAC Name
2,4-diisocyanato-1-methylbenzene
Molecular Formula
C9H6N2O2
InChI Key
DVKJHBMWWAPEIU-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1N=C=O)N=C=O
Chloride content
≤70 ppm (as hydrolyzable chloride)
Form
Clear liquid or solidified melt
Assay (GC)
18 to 22 wt% Toluene-2,6-diisocyanate content
Assay (GC)
78 to 82 wt% Toluene-2,4-diisocyanate content

Description

Toluene diisocyanate is utilized in polyurethane foams and tank trucks. It is also used in concrete sealers for aircraft, sealers and coatings in floor. It is employed as a crosslinking agent for nylon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text