Thermo Scientific Chemicals

Rosin, Thermo Scientific Chemicals

Catalog number: 036734.A1
1 kg, Each
Thermo Scientific Chemicals

Rosin, Thermo Scientific Chemicals

Catalog number: 036734.A1
1 kg, Each
Quantity
Catalog number: 036734.A1
also known as 036734-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8050-09-7
IUPAC Name
1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
Molecular Formula
C20H30O2
InChI Key
RSWGJHLUYNHPMX-UHFFFAOYNA-N
SMILES
CC(C)C1=CC2=CCC3C(C)(CCCC3(C)C(O)=O)C2CC1
Form
Solid
Saponification Value
170-220
Acid Value
160-190
Appearance (Color)
Cream to yellow to brown
Comment
Softening point: 70-80?C

Description

Rosin is an ingredient in printing inks, photocopying and laser printing paper, varnishes, adhesives (glues), soap, paper sizing, soda, soldering fluxes and sealing wax. It can be used as a glazing agent in medicines and chewing gum. In industry, rosin is a flux used in soldering. A mixture of pitch and rosin is used to make a surface against which glass is polished when making optical components such as lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text