Sodium metaborate tetrahydrate, tech., Thermo Scientific Chemicals
Sodium metaborate tetrahydrate, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium metaborate tetrahydrate, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number 036736.A7
also known as 036736-A7
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
Add to cart
Sodium metaborate tetrahydrate, tech., Thermo Scientific Chemicals
Catalog number036736.A7
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10555-76-7
IUPAC Namesodium tetrahydrate oxoborinate
Molecular FormulaBH8NaO6
InChI KeyJAKYJVJWXKRTSJ-UHFFFAOYSA-N
SMILESO.O.O.O.[Na+].[O-]B=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
Sodium metaborate tetrahydrate is used In textile industry, it is used as additives, process aid and flame retardant. In chemical industry, it is used to make boric acid. Other uses include thickening agent, preservative and photographic chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only