Thermo Scientific Chemicals

Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals

Catalog number: 036737.36
500 g, Each
Thermo Scientific Chemicals

Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals

Catalog number: 036737.36
500 g, Each
Quantity
Catalog number: 036737.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Form
Granules, 2-5 mm
Assay (special)
Suitability for moisture absorption: Passes Test (≥27.0% at 80% R.H.)

Description

Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text