Specifications
Chemical Name or MaterialGround & Waste Water Pollution Standard Solution 2
ConcentrationMatrix: 2% HNO3/tr. tartaric acid
Health Hazard 1H290-H315-H319
Quantity500 mL
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more