Thermo Scientific Chemicals
JM Special, Multi-element Oil Based Standard, Specpure™ 900μg/g, Thermo Scientific Chemicals
Catalog number: 036766.22
100 g, Each
Thermo Scientific Chemicals
JM Special, Multi-element Oil Based Standard, Specpure™ 900μg/g, Thermo Scientific Chemicals
Catalog number: 036766.22
100 g, Each
Quantity
Catalog number: 036766.22
also known as 036766-22
Price (USD)
Quantity
-
Have Questions?
Concentration
Mg 900?g/g ?2%
Concentration
Ca 900?g/g ?2%
Concentration
Zn 900?g/g ?2%
Concentration
Ba 900?g/g ?2%
Concentration
P 900?g/g ?2%