Thermo Scientific Chemicals

Boric acid, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 036771.36
500 g, Each
Thermo Scientific Chemicals

Boric acid, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 036771.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 036771.36
also known as 036771-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-35-3
IUPAC Name
boric acid
Molecular Formula
BH3O3
InChI Key
KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES
OB(O)O
Appearance (Color)
White
Form
Powder or crystals or granules
Total Metal Impurities
≤0.01%

Description

It is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is too employed in leather manufacture, electroplating, and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text