Thermo Scientific Chemicals

Phenylmercury acetate, 98%, Thermo Scientific Chemicals

Catalog number: 037125.30
250 g, Each
Thermo Scientific Chemicals

Phenylmercury acetate, 98%, Thermo Scientific Chemicals

Catalog number: 037125.30
250 g, Each
Quantity
Catalog number: 037125.30
also known as 037125-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
62-38-4
IUPAC Name
phenylmercurio acetate
Molecular Formula
C8H8HgO2
InChI Key
XEBWQGVWTUSTLN-UHFFFAOYSA-M
SMILES
CC(=O)O[Hg]C1=CC=CC=C1
Appearance (Color)
White to yellow
Assay (unspecified)
Polymercurated benzene compounds: 1.5%, max
Residue on ignition
0.2%, max
Assay (Titration)
> 97.5%
Form
crystalline powder

Description

Phenylmercuric acetate serves as contraceptive gel and foam, injectable solution, skin disinfectant and in cosmetic products such as hair shampoos, mouthwashes and toothpastes. It is likewise used in preservative in canned paint, eye creams and drops. It acts as a catalyst in polyurethane systems. It is also used as a catalyst in the 3M tartan brand polyurethane flexible floors that were common in school gymnasiums. It acts as an antitranspirant, when applied to the plants leaves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text