Glassy carbon rod, 1mm (0.04in) dia, type 2, Thermo Scientific Chemicals
Glassy carbon rod, 1mm (0.04in) dia, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon rod, 1mm (0.04in) dia, type 2, Thermo Scientific Chemicals

Widely used as a electrode material in electrochemistry.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mm
50 mm
200 mm
Catalog number 037996.DM
also known as 037996-DM
Price (USD)/ Each
495.65
Online exclusive
550.00 
Save 54.35 (10%)
-
Add to cart
Quantity:
100 mm
Request bulk or custom format
Price (USD)/ Each
495.65
Online exclusive
550.00 
Save 54.35 (10%)
Add to cart
Glassy carbon rod, 1mm (0.04in) dia, type 2, Thermo Scientific Chemicals
Catalog number037996.DM
Price (USD)/ Each
495.65
Online exclusive
550.00 
Save 54.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Diameter0.1 to 1.1 mm
Formsolid rod
Due to its distinctive property, it is widely used as a electrode material in electrochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only