Glassy carbon rod, 2mm (0.08in) dia, type 2, Thermo Scientific Chemicals
Glassy carbon rod, 2mm (0.08in) dia, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon rod, 2mm (0.08in) dia, type 2, Thermo Scientific Chemicals

Widely used as an electrode material in electrochemistry, as well as for high temperature crucibles and as a component of some prosthetic devices. It is used in laboratories as well.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mm
50 mm
200 mm
Catalog number 037997.DM
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
100 mm
Request bulk or custom format
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
Add to cart
Glassy carbon rod, 2mm (0.08in) dia, type 2, Thermo Scientific Chemicals
Catalog number037997.DM
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialGlassy carbon rod
Recommended StorageAmbient temperatures
EINECS Number231-153-3
TSCAYes
PackagingTube
View more
Glassy carbon rod, type 2 is widely used as an electrode material in electrochemistry, as well as for high temperature crucibles and as a component of some prosthetic devices. It is used in laboratories as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only