Glassy carbon rod, 5mm (0.2in) dia, type 2, Thermo Scientific Chemicals
Glassy carbon rod, 5mm (0.2in) dia, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon rod, 5mm (0.2in) dia, type 2, Thermo Scientific Chemicals

Widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mm
100 mm
200 mm
Catalog number 037998.DJ
also known as 037998-DJ
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
50 mm
Request bulk or custom format
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
Add to cart
Glassy carbon rod, 5mm (0.2in) dia, type 2, Thermo Scientific Chemicals
Catalog number037998.DJ
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 5mm (0.2in) diameter (Glassy Carbon type-2, maximimum service temperature 3000?C).
Appearance (Color)Black
Assay from Supplier's CofA>99.9%
Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only