Thermo Scientific Chemicals
Glassy carbon splinter powder, 80-200 micron, type 1, Thermo Scientific Chemicals
Catalog number: 038002.09
10 g, Each
Thermo Scientific Chemicals
Glassy carbon splinter powder, 80-200 micron, type 1, Thermo Scientific Chemicals
Catalog number: 038002.09
10 g, Each
Quantity
Catalog number: 038002.09
also known as 038002-09
Price (USD)
Quantity
-
Have Questions?
Form
Powder
Particle Size
80-200 micron: ≥90%
Description
Glassy carbon splinter powder is used as such in purifying metals, in electrodes, electrical devices & steel. L-Cysteine methyl ester modified glassy carbon powder (CysOMe-GC) has been shown to have potential for the removal of toxic heavy metal ions, such as copper(II), cadmium(II) and arsenic(III), from “real” water samples. Amorphous carbon may be used to develop zero temperature coefficient of resistance (TCR) based systems. Glassy carbon has been used as a catalyst support for bimetallic Rh-ReOx. It may be used to enhance the accuracy of infrared (IR) surface temperature measurements on micro-electro-mechanical systems (MEMS).