Glassy carbon splinter powder, 0.4 to 12μm, type 2, Thermo Scientific Chemicals
Glassy carbon splinter powder, 0.4 to 12μm, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon splinter powder, 0.4 to 12μm, type 2, Thermo Scientific Chemicals

Used as such in purifying metals, in electrodes, electrical devices & steel.
Have Questions?
Quantity:
50 g
Catalog number 038007.18
also known as 038007-18
Price (USD)/ Each
195.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
195.00
Add to cart
Glassy carbon splinter powder, 0.4 to 12μm, type 2, Thermo Scientific Chemicals
Catalog number038007.18
Price (USD)/ Each
195.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGlassy carbon splinter powder
Recommended StorageAmbient temperatures
TSCAYes
PackagingGlass bottle
MDL NumberMFCD00133992
View more

Glassy carbon splinter powder is used as such in purifying metals, in electrodes, electrical devices & steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only