Thermo Scientific Chemicals
Glassy carbon rod, 2mm (0.08in) dia, type 1, Thermo Scientific Chemicals
Catalog number: 038010.DR
200 mm, Each
Thermo Scientific Chemicals
Glassy carbon rod, 2mm (0.08in) dia, type 1, Thermo Scientific Chemicals
Catalog number: 038010.DR
200 mm, Each
Quantity
Catalog number: 038010.DR
also known as 038010-DR
Price (USD)
Quantity
-
Have Questions?
Diameter
1.8 to 2.2 mm
Appearance (Color)
Black
Form
Solid
Description
Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.